Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM166 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572215
Description
TMEM166 Polyclonal Antibody for Western Blot
Acts as a regulator of programmed cell death, mediating both autophagy and apoptosis. Expressed in lung, kidney, liver, pancreas, placenta, but not in heart and skeletal muscle.
Specifications
TMEM166 | |
Polyclonal | |
Unconjugated | |
EVA1A | |
BC014699; eva-1 homolog A; eva-1 homolog A (C. elegans); eva-1 homolog A, regulator of programmed cell death; EVA1A; fam176a; family with sequence similarity 176, member A; protein eva-1 homolog A; Protein FAM176A; RGD1559797; SP24; tm166; tmem166; transmembrane protein 166; zgc:153298 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
767647 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.66 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q08CB3 | |
EVA1A | |
KLH conjugated synthetic peptide between 104-137 amino acids from the Central region of Zebrafish THEM166. | |
400 μL | |
Primary | |
Zebrafish | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction