Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM166 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582331
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582331 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582331 Supplier Invitrogen™ Supplier No. PA582331
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ISCHTDCRRR PGKKFLQDRE SSSDSSDSED GSEDTVSDLS VRRHRRFERT LNKNVFTSAE ELERAQRLEE RERIIREIWM NGQPEVPGTR SLNRYY Highest antigen sequence indentity to the following orthologs: Rat - 92%, Mouse - 92%.

Acts as a regulator of programmed cell death, mediating both autophagy and apoptosis. Expressed in lung, kidney, liver, pancreas, placenta, but not in heart and skeletal muscle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM166
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EVA1A
Gene Accession No. Q9H8M9
Gene Alias BC014699; eva-1 homolog A; eva-1 homolog A (C. elegans); eva-1 homolog A, regulator of programmed cell death; EVA1A; fam176a; family with sequence similarity 176, member A; protein eva-1 homolog A; Protein FAM176A; RGD1559797; SP24; tm166; tmem166; transmembrane protein 166; zgc:153298
Gene Symbols EVA1A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EVA1A. Recombinant protein control fragment (Product #RP-90126).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84141
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.