Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM166 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596919
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596919 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596919 Supplier Invitrogen™ Supplier No. PA596919
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DTVSDLSVRR HRRFERTLNK NVFTSAEELE RAQRLEERER IIREIWMNGQ PEVPGTRSLN RYY Positive Samples: A-549, U-251MG, NCI-H460, Rat lung; Cellular Location: Endoplasmic reticulum membrane, Lysosome membrane, Single-pass membrane protein.

Acts as a regulator of programmed cell death, mediating both autophagy and apoptosis. Expressed in lung, kidney, liver, pancreas, placenta, but not in heart and skeletal muscle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM166
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.83 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene EVA1A
Gene Accession No. Q91WM6, Q9H8M9
Gene Alias BC014699; eva-1 homolog A; eva-1 homolog A (C. elegans); eva-1 homolog A, regulator of programmed cell death; EVA1A; fam176a; family with sequence similarity 176, member A; protein eva-1 homolog A; Protein FAM176A; RGD1559797; SP24; tm166; tmem166; transmembrane protein 166; zgc:153298
Gene Symbols EVA1A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 90-152 of human EVA1A (NP_1155571).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 232146, 500221, 84141
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.