Learn More
Invitrogen™ TMEM166 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113921
Description
Immunogen sequence: RLPLSHSPEH VEMALLSNIL AAYSFVSENP ERAALYFVSG VCIGLVLTLA For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Dog: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 79%; Rabbit: 93%.
Acts as a regulator of programmed cell death, mediating both autophagy and apoptosis. Expressed in lung, kidney, liver, pancreas, placenta, but not in heart and skeletal muscle.
Specifications
| TMEM166 | |
| Polyclonal | |
| Unconjugated | |
| EVA1A | |
| BC014699; eva-1 homolog A; eva-1 homolog A (C. elegans); eva-1 homolog A, regulator of programmed cell death; EVA1A; fam176a; family with sequence similarity 176, member A; protein eva-1 homolog A; Protein FAM176A; RGD1559797; SP24; tm166; tmem166; transmembrane protein 166; zgc:153298 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 84141 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9H8M9 | |
| EVA1A | |
| Synthetic peptide directed towards the N terminal region of human TMEM166 (aa 2-51). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.