Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM168 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554194
Description
Immunogen sequence: VLDSENSTPW VKEVRKINDQ YIAVQGAELI KTVDIEEADP PQLGDFTKDW VEYNCNSSNN ICWTEKGRTV K Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 93%.
TMEM168 is a multi-pass membrane protein. It belongs to the TMEM168 family. The exact function of TMEM168 remains unknown.
Specifications
| TMEM168 | |
| Polyclonal | |
| Unconjugated | |
| TMEM168 | |
| 5730526F17Rik; 8430437G11Rik; AI462344; AI504145; RGD1307778; Tmem168; Transmembrane protein 168 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64418 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H0V1 | |
| TMEM168 | |
| Recombinant protein corresponding to Human TMEM168. Recombinant protein control fragment (Product #RP-92749). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction