Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM169 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298157MFV500
Description
TMEM169 Polyclonal antibody specifically detects TMEM169 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
TMEM169 | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence | |
DKFZp781L2456, FLJ34263, transmembrane protein 169 | |
Produced in rabbits immunized with E. coli-derived Human TMEM169 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunocytochemistry | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
92691 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction