Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM169 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584550
Description
Immunogen sequence: VVQLSWSWEA WWQAARDMEK GFCGWLCSKL GLEDCSPYSI VELLESDNIS STLSNKDPIQ EVETST.
TMEM169 is a protein coding gene.
Specifications
TMEM169 | |
Polyclonal | |
Unconjugated | |
TMEM169 | |
A830020B06Rik; FLJ34263; hypothetical protein LOC617045; Tmem169; Transmembrane protein 169 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
92691 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96HH4 | |
TMEM169 | |
Recombinant protein corresponding to Human TMEM169. Recombinant protein control fragment (Product #RP-108472). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction