Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM16B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599001
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599001 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599001 Supplier Invitrogen™ Supplier No. PA599001
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TMEM16B Polyclonal Antibody for ELISA, ICC/IF, IHC (P), Western Blot

Calcium-activated chloride channels (CaCC) are present in many cell types and mediate physiological functions such as epithelial secretion, sensory signal transduction, and smooth muscle contraction. Subunits of these CaCC's include the transmembrane proteins TMEM16A and TMEM16B. TMEM16B is predicted to have eight transmembrane domains with both the amino and carboxy termini in the cytoplasm and is expressed in several tissues including olfactory sensory neurons as well as photoreceptors in mammalian retina. Like TMEM16A, TMEM16B is thought to form at least part of CaCC's but has different biophysical characteristics such as voltage dependence and unitary conductance.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM16B
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Ano2
Gene Accession No. Q9NQ90
Gene Alias ANO2; anoctamin 2; anoctamin 2 isoform Adelta4; anoctamin 2 isoform Bdelta4; anoctamin 2, calcium activated chloride channel; anoctamin 2-like; anoctamin-2; BC033409; C12orf3; LOC100361584; Tmem16b; Transmembrane protein 16B; transmembrane protein 16B (eight membrane-spanning domains)
Gene Symbols Ano2
Host Species Rabbit
Immunogen Recombinant Human Anoctamin-2 protein (1-83AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100361584, 57101
Target Species Human, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.