Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM16K Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543032
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543032 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543032 Supplier Invitrogen™ Supplier No. PA543032
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LKADIDATLY EQVILEKEMG TYLGTFDDYL ELFLQFGYVS LFSCVYPLAA Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.

TMEM16K is a multi-pass membrane protein. It belongs to the anoctamin family. TMEM16K may act as a calcium-activated chloride channel.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM16K
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Ano10
Gene Accession No. Q9NW15
Gene Alias AI604832; ANO10; ano10a; anoctamin 10; anoctamin 10a; anoctamin-10; fc37b04; hypothetical protein LOC566425; RGD1308260; SCAR10; Tmem16k; Transmembrane protein 16K; wu:fc37b04; zgc:114140
Gene Symbols Ano10
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human TMEM16K (aa 470-519).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55129
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.