Learn More
TMEM173/STING Rabbit, anti-Human, Unconjugated, Clone: 7K7, Proteintech

Rabbit Recombinant Monoclonal Antibody
$518.60
Specifications
Antigen | TMEM173/STING |
---|---|
Clone | 7K7 |
Concentration | 1000 μg/mL |
Applications | Western Blot, Immunohistochemistry, ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-225-6586
![]() ![]() |
Proteintech Group Inc
801651RR100UL |
100 μL |
Pack of 1 for $518.60
|
|
|||||
Description
TMEM173, also named as ERIS, MITA, STING and MPYS, is a facilitator of innate immune signaling that promotes the production of type I IFN (IFN-alpha and IFN-beta). TMEM173 mediates death signaling via activation of the extracellular signal-regulated kinase (ERK) pathway.
Specifications
TMEM173/STING | |
1000 μg/mL | |
Monoclonal | |
Liquid | |
Human, Mouse, Rat | |
Primary |
7K7 | |
Western Blot, Immunohistochemistry, ELISA | |
Unconjugated | |
Rabbit | |
340061 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.