Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM175 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100355
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100355 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100355 Supplier Invitrogen™ Supplier No. PA5100355
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TMEM175.

TMEM175 gene ontology annotations related to this gene include potassium channel activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM175
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TMEM175
Gene Accession No. Q6AY05, Q9BSA9, Q9CXY1
Gene Alias 3010001K23Rik; AI504381; endosomal/lysomomal potassium channel TMEM175; endosomal/lysosomal potassium channel TMEM175; hTMEM175; mTMEM175; RGD1309712; TMEM175; Transmembrane protein 175; transmembrane protein 175; endosomal/lysomomal potassium channel TMEM175
Gene Symbols TMEM175
Host Species Rabbit
Immunogen A synthesized peptide derived from human TMEM175(Accession Q9BSA9), corresponding to amino acid residues E262-A312.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 305623, 72392, 84286
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.