Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM175 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568634
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568634 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568634 Supplier Invitrogen™ Supplier No. PA568634
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Dog: 100%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 100%; Rat: 100%.

TMEM175 gene ontology annotations related to this gene include potassium channel activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM175
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TMEM175
Gene Accession No. Q9BSA9
Gene Alias 3010001K23Rik; AI504381; endosomal/lysomomal potassium channel TMEM175; endosomal/lysosomal potassium channel TMEM175; hTMEM175; mTMEM175; RGD1309712; TMEM175; Transmembrane protein 175; transmembrane protein 175; endosomal/lysomomal potassium channel TMEM175
Gene Symbols TMEM175
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human TMEM175.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84286
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.