Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM176A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542381
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542381 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542381 Supplier Invitrogen™ Supplier No. PA542381
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: GYSYYNSACR ISSSSDWNTP APTQSPEEVR RLHLCTSFMD MLKALFRTLQ Sequence homology: Horse: 79%; Human: 100%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM176A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TMEM176A
Gene Accession No. Q96HP8
Gene Alias 0610011i04rik; AU040201; AU041743; Gene signature 188; GS188; HCA112; Hepatocellular carcinoma-associated antigen 112; Keg2; kidney expressed gene 2; kidney-expressed gene 2 protein; likley ortholog of mouse GS188; MS4B1; RGD1310725; TMEM176A; Transmembrane protein 176A
Gene Symbols TMEM176A
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human TMEM176A.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55365
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.