Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM177 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571272
Description
This target displays homology in the following species: Dog: 86%; Guinea Pig: 86%; Horse: 79%; Human: 100%; Mouse: 86%; Rabbit: 79%; Rat: 100%.
Plays a role in the early steps of cytochrome c oxidase subunit II (MT-CO2/COX2) maturation and is required for the stabilization of COX20 and the newly synthesized MT-CO2/COX2 protein.
Specifications
TMEM177 | |
Polyclonal | |
Unconjugated | |
TMEM177 | |
2300008B03Rik; RGD1310320; TMEM177; Transmembrane protein 177 | |
Rabbit | |
Affinity chromatography | |
RUO | |
80775 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q53S58 | |
TMEM177 | |
synthetic peptide directed towards the C-terminal region of human TMEM177. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction