Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116855
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116855 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116855 Supplier Invitrogen™ Supplier No. PA5116855
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HeLa, A-549, Rat kidney Immunogen sequence: LLTCLSSRSY RLQIGHFLCL VILVYCAEYI NEAAAMNWRL FSKYQYFDSR GMFISIVFSA P.

Recent studies have examined how overexpression of certain genes enhances the tropism of neural stem cells (NSCs) for gliomas. Overexpression of Transmembrane protein 18 (TMEM18) provides NSCs and neural precursors an increased migration capacity towards glioblastoma cells in vitro and in the rat brain, while functional inactivation of the gene resulted in an almost complete loss of migration activity, suggesting that TMEM18 could be a potential target in NSC-based glioma therapy. Other recent results suggest that among others, the TMEM18 loci is associated with higher than normal body mass index (BMI) and obesity, indicating there may be a CNS-related role in predisposition to obesity. Multiple isoforms of TMEM18 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM18
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene TMEM18
Gene Accession No. Q6DGF8, Q96B42
Gene Alias BC024093; TMEM18; Transmembrane protein 18
Gene Symbols TMEM18
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 40-100 of human TMEM18 (NP_6900472).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 129787, 362722
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.