Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM18 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA520689
Description
A suggested positive control is rat brain tissue lysate. PA5-20689 can be used with blocking peptide PEP-0804.
Recent studies have examined how overexpression of certain genes enhances the tropism of neural stem cells (NSCs) for gliomas. Overexpression of Transmembrane protein 18 (TMEM18) provides NSCs and neural precursors an increased migration capacity towards glioblastoma cells in vitro and in the rat brain, while functional inactivation of the gene resulted in an almost complete loss of migration activity, suggesting that TMEM18 could be a potential target in NSC-based glioma therapy. Other recent results suggest that among others, the TMEM18 loci is associated with higher than normal body mass index (BMI) and obesity, indicating there may be a CNS-related role in predisposition to obesity. Multiple isoforms of TMEM18 are known to exist.
Specifications
| TMEM18 | |
| Polyclonal | |
| Unconjugated | |
| TMEM18 | |
| BC024093; TMEM18; Transmembrane protein 18 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 129787, 211986, 362722 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q3TUD9, Q6DGF8, Q96B42 | |
| TMEM18 | |
| A 15 amino acid peptide near the carboxy terminus of human TMEM18. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction