Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM181 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559097
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559097 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559097 Supplier Invitrogen™ Supplier No. PA559097
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KNALYESQLK DNPAFSMLND SDDDVIYGSD YEEMPLQNGQ AIRAKYKEES DSD Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

Mediates action of cytolethal distending toxins (CDT), which are secreted by many pathogenic bacteria. Expression level of TMEM181 is rate-limiting for intoxication.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM181
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMEM181
Gene Accession No. Q9P2C4
Gene Alias 5930418K15Rik; C76977; G protein-coupled receptor 178; GPR178; KIAA1423; mKIAA1423; TMEM181; Tmem181a; Transmembrane protein 181; transmembrane protein 181A
Gene Symbols TMEM181
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TMEM181. Recombinant protein control fragment (Product #RP-94899).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57583
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.