Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM182 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501734875 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4875 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4875 Supplier Proteintech Group Inc Supplier No. 253661AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TMEM182 is a protein coding gene.

Specifications

Antigen TMEM182
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene TMEM182
Gene Accession No. B2RVY9, Q6ZP80
Gene Alias TMEM182, transmembrane protein 182
Gene Symbols Tmem182
Host Species Rabbit
Immunogen TMEM182 Fusion Protein Ag21906
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 130827, 381339
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.