Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM184A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596834
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596834 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596834 Supplier Invitrogen™ Supplier No. PA596834
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FPCQVYAEKK ENSPAPPAPM QSISSGIRET VSPQDIVQDA IHNFSPAYQH YTQQATHEAP RPGTHPSGGS GGSRKSRSLE KRMLIPSEDL; Positive Samples: Mouse lung, Mouse liver, Mouse kidney, Rat lung; Cellular Location: Membrane, Multi-pass membrane protein.

TMEM184A, a multi-pass transmembrane protein, belongs to the UPF0206 family. While the exact function of TMEM184A remains unclear, it is known to be expressed at high levels in the developing testis. TMEM184A may mediate sex-specific signaling events during Sertoli cell differentiation. The restricted expression of this gene in Sertoli cells suggests a role for the encoded protein in the development and/or continued function of that cell type. Ectopic expression of TMEM184A in female genital ridges resulted in failure of gonocytes to enter meiosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM184A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.49 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TMEM184A
Gene Accession No. Q3UFJ6, Q4QQS1
Gene Alias RGD1306702; Sdmg1; Sexually dimorphic expressed in male gonads 1; tcag7.540; TMEM184A; Transmembrane protein 184A
Gene Symbols TMEM184A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 324-413 of human TMEM184A (NP_0010910891).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 231832, 304325
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.