Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM184A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564460
Description
Immunogen sequence: AYQHYTQQAT HEAPRPGTHP SGGSGGSRKS RSLEKRMLIP SE Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 63%.
TMEM184A, a multi-pass transmembrane protein, belongs to the UPF0206 family. While the exact function of TMEM184A remains unclear, it is known to be expressed at high levels in the developing testis. TMEM184A may mediate sex-specific signaling events during Sertoli cell differentiation. The restricted expression of this gene in Sertoli cells suggests a role for the encoded protein in the development and/or continued function of that cell type. Ectopic expression of TMEM184A in female genital ridges resulted in failure of gonocytes to enter meiosis.
Specifications
TMEM184A | |
Polyclonal | |
Unconjugated | |
TMEM184A | |
RGD1306702; Sdmg1; Sexually dimorphic expressed in male gonads 1; tcag7.540; TMEM184A; Transmembrane protein 184A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
202915 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZMB5 | |
TMEM184A | |
Recombinant protein corresponding to Human TMEM184A. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction