Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM184C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562896
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562896 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562896 Supplier Invitrogen™ Supplier No. PA562896
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSSSSQDAIS IASSMPPSPM GHYQGFGHTV TPQTTPTTAK ISDEILSDTI GEKKEPSDKS VD Highest antigen sequence identity to the following orthologs: Mouse - 45%, Rat - 40%.

Anaplastic thyroid cancer is one of the most lethal forms of cancer, but the precise carcinogenic mechanism has not been identified. TMEM184C, also known as TMEM34, was identified in a cDNA microarray analysis as being down-regulated in anaplastic thyroid cancers compared to normal thyroid tissues. TMEM184C protein expression was also lower in cell lines derived from these types of cancers compared to that of normal thyroid tissues or cell lines based on other types of thyroid cancers. Furthermore, transfection of TMEM34 into KTA2 cells led to the inhibition of cell growth, suggesting that TMEM184C might act as a tumor suppressor in anaplastic thyroid cancers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM184C
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMEM184C
Gene Accession No. Q9NVA4
Gene Alias 8430433H16Rik; AI645453; AL033298; BC004056; DNA sequence AY228474; PRO1355; TMEM184C; Tmem34; Transmembrane protein 184C; transmembrane protein 34
Gene Symbols TMEM184C
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TMEM184C. Recombinant protein control fragment (Product #RP-101430).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55751
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.