Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM185A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP25777325UL
Description
TMEM185A Polyclonal specifically detects TMEM185A in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
TMEM185A | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
chromosome X open reading frame 13, CXorf13, ee3, FAM11AMGC118845, family with sequence similarity 11, member A, fragile site, folic acid type, rare, fra(X)(q28) F, FRAXF, MGC118844, Protein FAM11A, transmembrane protein 185A | |
Rabbit | |
Affinity Purified | |
RUO | |
84548 | |
Human | |
IgG |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide | |
TMEM185A | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:AACVWGFRHDRSLELEILCSV | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction