Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM185A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561703
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561703 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561703 Supplier Invitrogen™ Supplier No. PA561703
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VGTGVWARNP QYRAEGETCV EFK Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

The protein encoded by this gene is predicted to be a transmembrane protein. This gene is best known for localizing to the CpG island of the fragile site FRAXF. The 5' untranslated region of this gene contains a CGG trinucleotide repeat sequence that normally consists of 7-40 tandem CGG repeats but which can expand to greater than 300 repeats. Methylation of the CpG island leads to transcriptional silencing of this gene, but neither the silencing nor an expanded repeat region appear to manifest itself in a clear phenotypic manner. Alternative splicing results in multiple transcript variants. A pseudogene of this gene has been defined on the X chromosome. [provided by RefSeq, Aug 2013].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM185A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMEM185A
Gene Accession No. Q8NFB2
Gene Alias BC023829; cDNA sequence BC023829; CXorf13; Ee3; erythropoietin-induced EST 3; FAM11A; family with sequence similarity 11, member A; fragile site, folic acid type, rare, fra(X)(q28) F; FRAXF; Protein FAM11A; Tmem185a; transmembrane protein 185A
Gene Symbols TMEM185A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TMEM185A. Recombinant protein control fragment (Product #RP-100167).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84548
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.