Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM186 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112753
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112753 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112753 Supplier Invitrogen™ Supplier No. PA5112753
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TMEM186 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

This gene encodes a potential transmembrane protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM186
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene TMEM186
Gene Accession No. Q96B77, Q9CR76
Gene Alias 2810440M13Rik; 4432406C05Rik; AI648175; AW210586; C16orf51; TMEM186; Transmembrane protein 186
Gene Symbols TMEM186
Host Species Rabbit
Immunogen Recombinant Human Transmembrane protein 186 protein (124-213AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25880, 66690
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.