Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM187 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569375
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569375 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569375 Supplier Invitrogen™ Supplier No. PA569375
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Dog: 79%; Horse: 77%; Human: 100%; Rabbit: 86%.

This gene consists of two exons and encodes a multi-pass membrane protein. An alternatively spliced transcript variant encoding the same protein has been found, but its biological validity is not determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM187
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TMEM187
Gene Accession No. Q14656
Gene Alias CXorf12; DXS9878E; ITBA1; Protein ITBA1; TMEM187; Transmembrane protein 187
Gene Symbols TMEM187
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human TMEM187.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8269
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.