Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM192 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598197
Description
TMEM192 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP, ELISA
TMEM192 is a widely expressed, four transmembrane protein that localizes to late endosomal/lysosomal membranes. This localization is mediated by two dileucine motifs present at the amino-terminus of TMEM192. A cysteine residue near the carboxy residue is responsible for disulfide bond formation in TMEM192 homodimers. Little is known about the function of this protein.
Specifications
TMEM192 | |
Polyclonal | |
Unconjugated | |
TMEM192 | |
3110005G23Rik; FLJ38482; RGD1309341; TMEM192; Transmembrane protein 192 | |
Rabbit | |
Protein G | |
RUO | |
201931 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry, Western Blot | |
2 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q8IY95 | |
TMEM192 | |
Recombinant Human Transmembrane protein 192 protein (193-271AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction