Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM192 Recombinant Rabbit Monoclonal Antibody (21G8)
GREENER_CHOICE

Catalog No. FEMA550066
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550066 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550066 Supplier Invitrogen™ Supplier No. MA550066
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

TMEM192 Recombinant Monoclonal Antibody for Western Blot, IHC (P), ELISA

TMEM192 is a widely expressed, four transmembrane protein that localizes to late endosomal/lysosomal membranes. This localization is mediated by two dileucine motifs present at the amino-terminus of TMEM192. A cysteine residue near the carboxy residue is responsible for disulfide bond formation in TMEM192 homodimers. Little is known about the function of this protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM192
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 21G8
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TMEM192
Gene Accession No. Q8IY95
Gene Alias 3110005G23Rik; FLJ38482; RGD1309341; TMEM192; Transmembrane protein 192
Gene Symbols TMEM192
Host Species Rabbit
Immunogen A synthesized peptide derived from human TMEM192.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201931
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.