Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM194A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546647
Description
Peptide sequence: LSGCLVYGTA ETDVNVVMLQ ESQVCEKRAS QQFCYTNVLI PKWHDIWTRI Sequence homology: Human: 100%.
The function of this protein remains unknown.
Specifications
TMEM194A | |
Polyclonal | |
Unconjugated | |
NEMP1 | |
Kiaa0286; NEMP1; Nuclear envelope integral membrane protein 1; Tmem194; Tmem194a; transmembrane protein 194; transmembrane protein 194A | |
Rabbit | |
Affinity Chromatography | |
RUO | |
23306 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O14524 | |
NEMP1 | |
Synthetic peptide directed towards the N-terminal of human TMEM194A (aa 34-83). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction