Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM194A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553157
Description
Immunogen sequence: LCTKNLEHPI QWLYITCRKV CKGAEKPVPP RLLTEEEYRI QGEVETRKAL EELREFCNSP DCSAWKTVSR IQSPKRFADF VEGSSHLTPN EVSVHEQEYG LGSIIAQDEI YEEASSEEED SYSRCPAITQ NNF Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 80%.
The function of this protein remains unknown.
Specifications
| TMEM194A | |
| Polyclonal | |
| Unconjugated | |
| NEMP1 | |
| Kiaa0286; NEMP1; Nuclear envelope integral membrane protein 1; Tmem194; Tmem194a; transmembrane protein 194; transmembrane protein 194A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23306 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O14524 | |
| NEMP1 | |
| Recombinant protein corresponding to Human TMEM194A. Recombinant protein control fragment (Product #RP-91173). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction