Learn More
Invitrogen™ TMEM198 Polyclonal Antibody

Description
Immunogen sequence: GTVATLRFQLL PPEPDDAFWG APCEQPLERR YQ Highest antigen sequence identity to the following orthologs - mouse 91%, rat 88%.
Specifications
Specifications
| Antigen | TMEM198 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | TMEM198 |
| Gene Accession No. | Q66K66 |
| Gene Alias | TMEM198; TMEM198A; Transmembrane protein 198 |
| Gene Symbols | TMEM198 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.