Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM199 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306494PE
Description
TMEM199 Polyclonal antibody specifically detects TMEM199 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
TMEM199 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
C17orf32, chromosome 17 open reading frame 32, MGC45714, transmembrane protein 199 | |
Produced in rabbits immunized with E. coli-derived Human TMEM199 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunoprecipitation | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
147007 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction