Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM199 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555496
Description
Immunogen sequence: AGERLVRALG PGGELEPERL PRKLRAELEA ALGKKHKGGD SSSGPQRLVS FRLIRDLHQH LRERDSKLYL HELLEGSEIY LPEVVKPPRN PELVARLEKI KIQLANEEYK RITRNVTCQD TRHGGTLSDL GKQVRSL Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.
The function of this protein remains unknown.
Specifications
| TMEM199 | |
| Polyclonal | |
| Unconjugated | |
| TMEM199 | |
| AI316787; AI848469; C17orf32; CDG2P; hypothetical protein LOC553562; R74819; RE40914p; TMEM199; Transmembrane protein 199; Unknown (protein for MGC:137613); VMA12; VPH2; zgc:109974 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 147007, 195040, 303332 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5BK13, Q5SYH2, Q8N511 | |
| TMEM199 | |
| Recombinant protein corresponding to Human TMEM199. Recombinant protein control fragment (Product #RP-91003). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction