Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5141157
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5141157 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5141157 Supplier Invitrogen™ Supplier No. PA5141157
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TMEM2 Polyclonal Antibody for Western Blot

This gene encodes a type II transmembrane protein that belongs to the interferon-induced transmembrane (IFITM) protein superfamily. The encoded protein functions as a cell surface hyaluronidase that cleaves extracellular high molecular weight hyaluronan into intermediate size fragments before internalization and degradation in the lysosome. It also has an interferon-mediated antiviral function in humans through activation of the JAK STAT signaling pathway. The activation of this gene by transcription factor SOX4 in breast cancer cells has been shown to mediate the pathological effects of SOX4 on cancer progression. Naturally occurring mutations in this gene are associated with autosomal recessive non-syndromic hearing loss.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene CEMIP2
Gene Accession No. Q5FWI3, Q9UHN6
Gene Alias basic helix-loop-helix domain containing, class B, 2; BHLHB2; cell migration inducing hyaluronidase 2; Cell migration-inducing hyaluronidase 2; Cell surface hyaluronidase; CEMIP2; Kiaa1412; Tmem2; transmembrane protein 2
Gene Symbols CEMIP2
Host Species Rabbit
Immunogen Synthetic peptide within Human TMEM2 aa 98-147.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23670, 309400, 83921
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.