Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM200A Monoclonal Antibody (4-B3)
GREENER_CHOICE

Catalog No. PIMA532881
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532881 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532881 Supplier Invitrogen™ Supplier No. MA532881
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TMEM200A Monoclonal Antibody for ICC/IF, IHC (P), Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM200A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 4-B3
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% Glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4
Gene TMEM200A
Gene Accession No. Q86VY9, Q8C817
Gene Alias C030003D03Rik; HBE61; Kiaa1913; Tmem200a; Transmembrane protein 200A; TTMA; TTMC; two transmembrane C
Gene Symbols TMEM200A
Host Species Mouse
Immunogen Recombinant protein within Human Transmembrane protein 200A aa 59-359.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114801, 77220
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.