Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM200A Mouse anti-Human, Clone: 4-B3, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP267214
Description
TMEM200A Monoclonal specifically detects TMEM200A in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TMEM200A | |
Monoclonal | |
2 mg/mL | |
Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:50 | |
found in a patient with schizophrenia, KIAA1913TTMC, transmembrane protein 200A, TTMA, two transmembrane C | |
Mouse | |
Immunogen affinity purified | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
4-B3 | |
Unconjugated | |
PBS (pH7.4), 0.2% BSA, 40% Glycerol with 0.05% Sodium Azide | |
TMEM200A | |
Recombinant protein within Human TMEM200A aa 59-359/ 491. (SwissProt: Q86VY9 Human; SwissProt: Q8C817 Mouse) | |
100 μL | |
114801 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction