Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM200C Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561480
Description
Immunogen sequence: SSQSDDPSSS NKGYTPLREA GTSTESVLDA VAGQTRDSAV AAPVLGAEQS SPEGASQEPP TAEQPQPVQ Highest antigen sequence identity to the following orthologs: Mouse - 61%, Rat - 58%.

Specifications
TMEM200C | |
Polyclonal | |
Unconjugated | |
TMEM200C | |
TMEM200C; Transmembrane protein 200C; transmembrane protein TTMA; TTMA; two transmembrane domain family member A; two transmembrane domain-containing family member A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
645369 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6NKL6 | |
TMEM200C | |
Recombinant protein corresponding to Human TMEM200C. Recombinant protein control fragment (Product #RP-100785). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction