Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM204 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553112
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553112 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553112 Supplier Invitrogen™ Supplier No. PA553112
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HKREDCMAPR VIVISRSLTA RFRRGLDNDY VESPC Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

Transmembrane protein 204 (TMEM204) is a four-transmembrane protein that is induced under hypoxic conditions. It localizes to cell-cell junctions and co-localizes with the beta-catenin adherens junction-associated protein, but not with tight junctions. Overexpression of TMEM204 results in decreased adhesion between cells, indicating that TMEM204 is an adherens junction protein that modulates cell adhesion and potentially angiogenesis. Recent evidence suggests TMEM204 may also be involved in epilepsy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM204
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMEM204
Gene Accession No. Q9BSN7
Gene Alias C16orf30; Claudin-like protein 24; claudin-like protein of 24 kDa; Clp24; Tmem204; Transmembrane protein 204; UNQ6509/PRO21434
Gene Symbols TMEM204
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TMEM204. Recombinant protein control fragment (Product #RP-90756).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79652
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.