Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM205 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA519405
Description
This antibody is predicted to react with canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.

Specifications
TMEM205 | |
Polyclonal | |
Unconjugated | |
TMEM205 | |
MBC3205; MGC110858; MGC3205; TMEM205; Transmembrane protein 205; UNQ501; UNQ501/PRO1018 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
374882 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q6UW68 | |
TMEM205 | |
Synthetic peptide sequence (PDPYRQLREK) corresponding to the internal amino acids of TMEM205 (aa 137-147). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction