Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM206 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA552575
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552575 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552575 Supplier Thermo Scientific Supplier No. PA552575

Rabbit Polyclonal Antibody

Immunogen sequence: LKHPVMSVSY KEVDRYDAPG IALYPGQAQL LSCKHHYEVI PPLTSPGQPG DMNCTTQRIN YTDPFSNQTV KSALIVQGPR EVKKRELVFL QFRLNKSSED FSAIDYLLFS SFQEFLQSPN RV Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 89%.

TMEM206 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM206
Applications Immunohistochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMEM206
Gene Accession No. Q9H813
Gene Alias 2310028N02Rik; AI118576; C16H1orf75; C1orf75; MGC137693 protein; PAC; PACC1; proton activated chloride channel 1; proton-activated chloride channel; RGD1359339; Tmem206; transmembrane protein 206; Transmembrane protein 206-like protein
Gene Symbols TMEM206
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TMEM206
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55248
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.