Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM208 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559800
Description
Immunogen sequence: PWFTADSGTP APEHNEKRQR RQERRQMKRL Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 90%.
May function as a negative regulator of endoplasmic reticulum-stress induced autophagy.
Specifications
TMEM208 | |
Polyclonal | |
Unconjugated | |
TMEM208 | |
1700006C06Rik; 2610030K20Rik; HSPC171; hypothetical protein zgc:77041; TMEM208; Transmembrane protein 208; zgc:77041 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
29100 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BTX3 | |
TMEM208 | |
Recombinant protein corresponding to Human TMEM208. Recombinant protein control fragment (Product #RP-98757). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction