Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM208 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559764
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559764 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559764 Supplier Invitrogen™ Supplier No. PA559764
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SYHSMSSMAR AAFSEDGALM DGGMDLNMEQ GMAEHLKDV Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 92%.

May function as a negative regulator of endoplasmic reticulum-stress induced autophagy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM208
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMEM208
Gene Accession No. Q9BTX3
Gene Alias 1700006C06Rik; 2610030K20Rik; HSPC171; hypothetical protein zgc:77041; TMEM208; Transmembrane protein 208; zgc:77041
Gene Symbols TMEM208
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TMEM208. Recombinant protein control fragment (Product #RP-98758).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29100
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.