Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM209 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556818
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556818 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556818 Supplier Invitrogen™ Supplier No. PA556818
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: APCANKDEAD LSSKQAAEEV WARVAMNRQL LDHMDSWTAK FRNWINETIL VPLVQEIESV STQMRRMGCP ELQIGEASIT SLKQAALVKA PLIPTLNTIV QYLDLTPNQE YLFERIKELS QGGCMSSFRW NRGGDFKGRK WDTD Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

TMEM209 (Transmembrane Protein 209) is a Protein Coding gene. Diseases associated with TMEM209 include Hyperalphalipoproteinemia 1. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM209
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMEM209
Gene Accession No. Q96SK2
Gene Alias 2700094F01Rik; AI428435; NET31; RGD1309682; testicular tissue protein Li 202; Tmem209; Transmembrane protein 209; Transmembrane protein 209-like protein
Gene Symbols TMEM209
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TMEM209. Recombinant protein control fragment (Product #RP-90987).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84928
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.