Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM212 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565415
Description
Immunogen sequence: VTFPYPYAKFP LACVDPPHYE EYHLTLQ Highest antigen sequence identity to the following orthologs - mouse 82%, rat 79%.
TMEM212 is a protein coding gene.
Specifications
TMEM212 | |
Polyclonal | |
Unconjugated | |
TMEM212 | |
TMEM212; Transmembrane protein 212 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
389177 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6NML5 | |
TMEM212 | |
Recombinant Human TMEM212. Recombinant protein control fragment (Product #RP-100465). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction