Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM214 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB183848 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB183848 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB183848 Supplier Novus Biologicals Supplier No. NBP282034AF350
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TMEM214 Polyclonal antibody specifically detects TMEM214 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen TMEM214
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias FLJ20254, FLJ39682, PP446, transmembrane protein 214
Host Species Rabbit
Immunogen TMEM214 antibody was raised against a 17 amino acid synthetic peptide near the center of human TMEM214. The immunogen is located within amino acids 150 - 200 of TMEM214. Amino Acid Squence: HTHDYPYSLVSRELRGI
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54867
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.