Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM215 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584588
Description
Immunogen sequence: TEGCTKWPEN ELLWVRKLPC FRKPKDKEVV ELLRTPSDLE SGKGSSDELA KKAGLRGKPP PQSQGE.
TMEM215 is a protein coding gene.
Specifications
TMEM215 | |
Polyclonal | |
Unconjugated | |
TMEM215 | |
A930001M12Rik; hypothetical protein LOC690918; LOC690918; TMEM215; Transmembrane protein 215 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
401498 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q68D42 | |
TMEM215 | |
Recombinant protein corresponding to Human TMEM215. Recombinant protein control fragment (Product #RP-108484). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction