Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM217 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$377.03
Specifications
| Antigen | TMEM217 |
|---|---|
| Dilution | Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
TMEM217 Polyclonal antibody specifically detects TMEM217 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| TMEM217 | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| C6orf128, Chromosome 6 Open Reading Frame 128, DJ355M6.2, Transmembrane Protein 217 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: SQGNIISYKRRISTAEILHSRNKRLSISSGFSGSHLESQYFERQSFHTSIFTCLSPVPSSAPSTCRYTIDV | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol | |
| 221468 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title