Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM217 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA584859

Catalog No. PIPA584859


Only null left
Add to Cart

Description

Description

Immunogen sequence: VIQILTNNDF DIKEVRIMRW FGLV.

TMEM217 is a protein coding gene. Gene ontology (GO) annotation include fibrillar center; integral component of membrane; nucleolus.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TMEM217
Polyclonal
Unconjugated
TMEM217
C6orf128; dJ355M6.2; TMEM217; Transmembrane protein 217
Rabbit
Antigen Affinity Chromatography
RUO
221468
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunofluorescence
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8N7C4
TMEM217
Recombinant protein corresponding to Human TMEM217. Recombinant protein control fragment (Product #RP-108580).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.