Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM229A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563294
Description
Immunogen sequence: SPDLRMLGFS SPYRCLLHSL THFALEKVYL QQRRCPNAFV FNFLLYPSAH VGLQTLAGQA LLLSLGG Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.

Specifications
TMEM229A | |
Polyclonal | |
Unconjugated | |
TMEM229A | |
6332401O19Rik; tcag7.977; Tmem229a; Transmembrane protein 229A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
730130 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
B2RXF0 | |
TMEM229A | |
Recombinant protein corresponding to Human TMEM229A. Recombinant protein control fragment (Product #RP-101802). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction