Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM230 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566391
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566391 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566391 Supplier Invitrogen™ Supplier No. PA566391
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LRIAYYASKGY RGYSYDDIPD FDD Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

Involved in trafficking and recycling of synaptic vesicles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM230
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMEM230
Gene Accession No. Q96A57
Gene Alias 5730494N06Rik; AA407821; C20orf30; dJ1116H23.2.1; HSPC274; HSPC274 protein; RGD1307399; TMEM230; Transmembrane protein 230; UNQ2432/PRO4992; UPF0414 transmembrane protein C20orf30; UPF0414 transmembrane protein C20orf30 homolog
Gene Symbols TMEM230
Host Species Rabbit
Immunogen Recombinant Human TMEM230. Recombinant protein control fragment (Product #RP-105186).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29058
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.