Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM231 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542686
Description
Peptide sequence: CHEAPRARSA RAGLPNRLPT ALFNSGFWLK RSSYEEQPTV RFQHQVLLVA Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%.
The function remains unknown.
Specifications
TMEM231 | |
Polyclonal | |
Unconjugated | |
TMEM231 | |
4932417I16Rik; ALYE870; carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 5; chst5; Gm178; JBTS20; MKS11; PRO1886; RGD1306153; tmem231; Transmembrane protein 231; UNQ870/PRO1886; zgc:64166 | |
Rabbit | |
Protein A | |
RUO | |
79583 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9H6L2 | |
TMEM231 | |
Synthetic peptide directed towards the N-terminal of human FLJ22167. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction